Assay Details
Target Gene Details
Entrez Gene ID: | 14677 |
Gene Name: | guanine nucleotide binding protein (G protein), alpha inhibiting 1 |
Gene Aliases: |
AU046200, Gialpha1, Gnai-1 |
Location: |
Chr.5:18265135-18360413 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnai1 | NM_010305.1 | 1 | 235 | NP_034435.1 |
BC138862.1 | 1 | 115 | ||
BC138865.1 | 1 | 115 | ||
BY740128.1 | 1 | 235 |
More Information
Set Membership: |
Intragenic Exonic |