Assay Details
Target Gene Details
Entrez Gene ID: | 22062 |
Gene Name: | transformation related protein 73 |
Gene Aliases: |
TAp73, Tp73, deltaNp73, p73 |
Location: |
Chr.4:154056249-154140208 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trp73 | NM_001126330.1 | 1 | 184 | NP_001119802.1 |
NM_001126331.1 | 1 | 184 | NP_001119803.1 | |
NM_011642.3 | NP_035772.2 | |||
AB221594.1 | ||||
AK139633.1 | 1 | 185 | BAE24089.1 | |
BB663805.1 | ||||
BC066045.1 | 1 | 71 | AAH66045.1 | |
Y19234.1 | CAB81953.1 | |||
Y19235.1 | 1 | 178 | CAB81954.1 |
More Information
Set Membership: |
Intragenic Exonic |