Assay Details
Target Gene Details
Entrez Gene ID: | 110208 |
Gene Name: | phosphogluconate dehydrogenase |
Gene Aliases: |
0610042A05Rik, AU019875, C78335 |
Location: |
Chr.4:149149985-149166707 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pgd | NM_001081274.1 | 13 | 1715 | NP_001074743.1 |
AK002894.1 | 13 | 1677 | BAB22439.1 | |
AK004217.1 | 6 | 917 | ||
AK077110.1 | 13 | 1674 | ||
AK145602.1 | 13 | 1679 | BAE26535.1 | |
AK150210.1 | 13 | 1678 | BAE29381.1 | |
AK150490.1 | 13 | 1674 | BAE29606.1 | |
AK151380.1 | 13 | 1674 | BAE30352.1 | |
AK151993.1 | 13 | 1674 | BAE30857.1 | |
AK152758.1 | 13 | 1674 | BAE31473.1 | |
AK152894.1 | 13 | 1674 | BAE31577.1 | |
AK153409.1 | 13 | 1671 | BAE31969.1 | |
AK155027.1 | 13 | 1674 | BAE32999.1 | |
AK166733.1 | 13 | 1669 | BAE38978.1 | |
AK166947.1 | 13 | 1651 | BAE39134.1 | |
AK167215.1 | 13 | 1669 | BAE39341.1 | |
AK168251.1 | 13 | 1674 | BAE40201.1 | |
BC008646.1 | 13 | 1646 | AAH08646.1 | |
BC011329.1 | 13 | 1664 | AAH11329.1 | |
BC014793.1 | 13 | 1483 | AAH14793.1 |
More Information
Set Membership: |
Intragenic Exonic |