Assay Details
Target Gene Details
Entrez Gene ID: | 67808 |
Gene Name: | transformation related protein 63 regulated like |
Gene Aliases: |
1200015A19Rik, Svap30, Tprg1l, mover |
Location: |
Chr.4:154157485-154160712 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tprgl | NM_026388.2 | 4 | 1570 | NP_080664.1 |
AI840915.1 | 1 | 187 | ||
AK004324.1 | 3 | 1183 | BAC25076.1 | |
AK004777.1 | 4 | 1553 | BAB23555.1 | |
AK155413.1 | 4 | 1565 | BAE33252.1 | |
AK183912.1 | 1 | 159 | ||
AK187964.1 | 1 | 160 | ||
AK198367.1 | 1 | 159 | ||
AK207759.1 | 1 | 159 | ||
AK208827.1 | 1 | 159 | ||
AK211533.1 | 1 | 162 | ||
AK211936.1 | 1 | 347 | ||
AK214783.1 | 1 | 159 | ||
AK216498.1 | 1 | 159 | ||
AK217635.1 | 1 | 159 | ||
AK218082.1 | 1 | 159 | ||
BC019544.1 | 4 | 1534 | AAH19544.1 |
Target Gene Details
Entrez Gene ID: | 59002 |
Gene Name: | WD repeat containing, antisense to Trp73 |
Gene Aliases: |
2610044M17Rik, 5330425N03Rik, Dd57, Wdr8 |
Location: |
Chr.4:154142357-154168259 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wrap73 | AK160273.1 | |||
AK172690.1 | BAE43131.1 |