Assay Details
Target Gene Details
Entrez Gene ID: | 231162 |
Gene Name: | cytokine-like 1 |
Gene Aliases: |
4930443F05Rik, 4Cytl1, C17, Cyt1, Gm147 |
Location: |
Chr.5:37735519-37739820 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cytl1 | NM_001081106.1 | 4 | 666 | NP_001074575.1 |
AK076770.1 | 3 | 558 | ||
BC063103.1 | 4 | 626 | AAH63103.1 |
More Information
Set Membership: |
Intragenic Exonic |