Assay Details
Target Gene Details
Entrez Gene ID: | 109979 |
Gene Name: | ADP-ribosyltransferase 3 |
Gene Aliases: |
4930569O04Rik, ARTC3 |
Location: |
Chr.5:92331799-92414628 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Art3 | NM_181728.2 | NP_859417.2 | ||
AK016257.1 | ||||
DQ519368.1 | ||||
DQ519369.1 | ||||
DQ810295.1 |
Target Gene Details
Entrez Gene ID: | 56066 |
Gene Name: | chemokine (C-X-C motif) ligand 11 |
Gene Aliases: |
Cxc11, H174, I-tac, Ip9, Itac, Scyb11, Scyb9b, b-R1, betaR1 |
Location: |
Chr.5:92359544-92363296 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl11 | NM_019494.1 | 4 | 744 | NP_062367.1 |
NR_038116.1 | 4 | 762 | ||
AF136449.1 | 4 | 744 | AAF79022.1 | |
AF178672.1 | 4 | 744 | AAF79048.1 | |
AF179872.1 | 4 | 744 | AAF77095.1 | |
AK040051.1 | 4 | 704 | ||
BC025903.1 | 4 | 732 | AAH25903.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |