Assay Details
Target Gene Details
Entrez Gene ID: | 12307 |
Gene Name: | calbindin 1 |
Gene Aliases: |
Brain-2, CB, Calb, Calb-1 |
Location: |
Chr.4:15881264-15906709 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Calb1 | NM_009788.4 | 11 | 2181 | NP_033918.1 |
AK002635.1 | 11 | 2183 | BAB22249.1 | |
AK028050.1 | 11 | 2182 | BAC25721.1 | |
AK028327.1 | 11 | 2180 | BAC25882.1 | |
AK042638.1 | 1 | 49 | ||
AK048517.1 | 11 | 2182 | BAC33354.1 |
More Information
Set Membership: |
Intragenic Exonic |