Assay Details
Target Gene Details
Entrez Gene ID: | 230126 |
Gene Name: | src homology 2 domain-containing transforming protein B |
Gene Aliases: |
BC028832 |
Location: |
Chr.4:45423276-45530828 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shb | NM_001033306.1 | 1 | 875 | NP_001028478.1 |
BC058986.1 | 1 | 377 | AAH58986.1 |