Assay Details
Target Gene Details
Entrez Gene ID: | 12457 |
Gene Name: | CCR4 carbon catabolite repression 4-like (S. cerevisiae) |
Gene Aliases: |
AU043840, Ccr4, nocturnin |
Location: |
Chr.3:51224447-51251654 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccrn4l | NM_009834.2 | 2 | 892 | NP_033964.1 |
AF183960.1 | 2 | 878 | AAD56547.1 | |
AF199491.1 | 2 | 385 | AAG01384.1 | |
AK041713.1 | 2 | 408 | ||
AK084516.1 | 2 | 413 | ||
AK087790.1 | 2 | 892 | BAC40004.1 | |
AK147356.1 | 1 | 2748 | ||
AK147433.1 | 1 | 2748 | ||
AK149340.1 | 2 | 870 | BAE28819.1 | |
BC139369.1 | 2 | 531 | ||
BC139370.1 | 2 | 531 | ||
U70139.1 | 2 | 7571 | AAB62717.1 |
More Information
Set Membership: |
Intragenic Exonic |