Assay Details
Target Gene Details
Entrez Gene ID: | 12457 |
Gene Name: | CCR4 carbon catabolite repression 4-like (S. cerevisiae) |
Gene Aliases: |
AU043840, Ccr4, nocturnin |
Location: |
Chr.3:51224447-51251654 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccrn4l | NM_009834.2 | 1 | 278 | NP_033964.1 |
AF183960.1 | 1 | 264 | AAD56547.1 | |
AK087790.1 | 1 | 278 | BAC40004.1 | |
AK149340.1 | 1 | 256 | BAE28819.1 |
More Information
Set Membership: |
Intragenic Exonic |