Assay Details
Target Gene Details
Entrez Gene ID: | 26570 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 11 |
Gene Aliases: |
9930009M05Rik, AI451155, sut, xCT |
Location: |
Chr.3:50364936-50499087 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a11 | NM_011990.2 | 1 | 219 | NP_036120.1 |
AB022345.1 | 1 | 186 | BAA77220.1 | |
AK037490.1 | 1 | 220 | ||
AK037742.1 | 1 | 194 | BAC29863.1 | |
AK076322.1 | 1 | 193 | ||
AK089202.1 | 1 | 192 | BAC40792.1 | |
AK089283.1 | 1 | 211 | ||
AK162233.1 | ||||
AK163168.1 | 1 | 193 | BAE37218.1 | |
AK165599.1 | 1 | 195 | BAE38284.1 | |
AK170568.1 | 1 | 88 | ||
AK171192.1 | 1 | 217 | BAE42302.1 | |
AK171252.1 | 1 | 193 | BAE42344.1 | |
AK171327.1 | 1 | 195 | ||
AY766236.1 | 1 | 186 | AAX10866.1 | |
BB630675.1 | 1 | 220 | ||
BC141402.1 | 1 | 116 | ||
BC141408.1 | 1 | 116 |
More Information
Set Membership: |
Intragenic Exonic |