Assay Details
Target Gene Details
Entrez Gene ID: | 18160 |
Gene Name: | natriuretic peptide receptor 1 |
Gene Aliases: |
AI893888, GC-A, NPR-A, NPRA, Pndr |
Location: |
Chr.3:90450592-90465928 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npr1 | NM_008727.5 | 22 | 3871 | NP_032753.5 |
AK135008.1 | 13 | 1814 | BAE22383.1 | |
BC110659.1 | 22 | 3751 | AAI10660.1 | |
J05504.1 | 23 | 3855 | AAA37670.1 |
More Information
Set Membership: |
Intragenic Exonic |