Assay Details
Target Gene Details
Entrez Gene ID: | 72413 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 2 |
Gene Aliases: |
2700049B16Rik, 3110031N04Rik |
Location: |
Chr.3:31902132-32200180 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnmb2 | NM_028231.2 | 6 | 1157 | NP_082507.1 |
AB193520.1 | ||||
AB193521.1 | ||||
AK012400.1 | 6 | 1008 | BAB28216.1 | |
AK014106.1 | 5 | 1253 | ||
AY062429.1 | AAL38982.1 | |||
BC046227.1 | 6 | 1157 | AAH46227.1 | |
BC058957.1 | 6 | 1157 | AAH58957.1 |
More Information
Set Membership: |
Intragenic Exonic |