Assay Details
Target Gene Details
Entrez Gene ID: | 76080 |
Gene Name: | tocopherol (alpha) transfer protein-like |
Gene Aliases: |
3110080A02Rik, 5830472M02Rik |
Location: |
Chr.2:163602281-163619013 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttpal | NM_029512.2 | 5 | 3295 | NP_083788.2 |
NM_181734.3 | 5 | 3292 | NP_859423.2 | |
AK014262.1 | 1 | 637 | ||
AK156641.1 | 5 | 3258 | BAE33788.1 | |
BC052389.1 | 5 | 3251 |
More Information
Set Membership: |
Intragenic Exonic |