Assay Details
Target Gene Details
Entrez Gene ID: | 20197 |
Gene Name: | S100 calcium binding protein A3 |
Gene Aliases: |
S100E |
Location: |
Chr.3:90560356-90602706 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100a3 | NM_011310.2 | 3 | 289 | NP_035440.1 |
AF004941.1 | 3 | 289 | AAC78730.1 | |
AF087470.1 | 3 | 264 | AAC64107.1 | |
AK133717.1 | 3 | 291 | BAE21797.1 | |
AK161105.1 | 1 | 2036 | ||
BC099893.1 | 3 | 286 | AAH99893.1 | |
BC100690.1 | 3 | 286 | AAI00691.1 | |
BC100696.1 | 3 | 286 | AAI00697.1 | |
BC100697.1 | 3 | 286 | AAI00698.1 |
More Information
Set Membership: |
Intragenic Exonic |