Assay Details
Target Gene Details
Entrez Gene ID: | 66634 |
Gene Name: | minichromosome maintenance deficient 8 (S. cerevisiae) |
Gene Aliases: |
5730432L01Rik |
Location: |
Chr.2:132816141-132844197 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcm8 | NM_001291054.1 | 19 | 2877 | NP_001277983.1 |
NM_025676.4 | 18 | 2793 | NP_079952.2 | |
AK010365.1 | 10 | 1515 | BAB26885.1 | |
AK017607.1 | 19 | 2774 | ||
AK133858.1 | 19 | 2834 | BAE21892.1 | |
BC046780.1 | 19 | 2824 | AAH46780.1 | |
BC052070.1 | 18 | 2599 | AAH52070.1 |
More Information
Set Membership: |
Intragenic Exonic |