Assay Details
Target Gene Details
Entrez Gene ID: | 240726 |
Gene Name: | solute carrier organic anion transporter family, member 5A1 |
Gene Aliases: |
4732454B15, A630033C23Rik, AA407151 |
Location: |
Chr.1:12866549-12992932 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slco5a1 | NM_172841.2 | 9 | 7545 | NP_766429.2 |
AK149216.1 | 1 | 1308 |
More Information
Set Membership: |
Intragenic Exonic |