Assay Details
Target Gene Details
Entrez Gene ID: | 22637 |
Gene Name: | zeta-chain (TCR) associated protein kinase |
Gene Aliases: |
Srk, ZAP-70, mrtle, mur |
Location: |
Chr.1:36761798-36782821 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zap70 | NM_001289612.1 | 6 | 1217 | NP_001276541.1 |
NM_001289765.1 | 13 | 2164 | NP_001276694.1 | |
NM_001289766.1 | 13 | 2129 | NP_001276695.1 | |
NM_009539.3 | 13 | 2121 | NP_033565.2 | |
AB083210.1 | 6 | 1220 | BAC43746.1 | |
AK039883.1 | 13 | 2058 | BAC30471.1 | |
AK089056.1 | 13 | 2093 | ||
BC029727.1 | 13 | 2079 | AAH29727.1 | |
BY589032.1 | 1 | 71 | ||
U04379.1 | AAA19250.1 |