Assay Details
Target Gene Details
Entrez Gene ID: | 241296 |
Gene Name: | leucine rich repeat containing 8A |
Gene Aliases: |
Lrrc8, mKIAA1437 |
Location: |
Chr.2:30237769-30263790 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrc8a | NM_177725.4 | 4 | 2957 | NP_808393.1 |