Assay Details
Target Gene Details
Entrez Gene ID: | 497097 |
Gene Name: | X Kell blood group precursor related family member 4 |
Gene Aliases: |
AY534250, Gm210, XRG4, mKIAA1889 |
Location: |
Chr.1:3199731-3671742 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Xkr4 | NM_001011874.1 | 3 | 3229 | NP_001011874.1 |
AK135172.1 | 1 | 746 | ||
AK220428.1 | 1 | 1458 | BAD90263.1 | |
AY534250.1 | 3 | 3229 | AAT07099.1 |