Assay Details
Target Gene Details
Entrez Gene ID: | 11350 |
Gene Name: | c-abl oncogene 1, non-receptor tyrosine kinase |
Gene Aliases: |
AI325092, Abl, E430008G22Rik, c-Abl |
Location: |
Chr.2:31688252-31807093 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abl1 | NM_001112703.2 | 4 | 1962 | NP_001106174.1 |
NM_001283045.1 | 4 | 839 | NP_001269974.1 | |
NM_001283046.1 | 5 | 2167 | NP_001269975.1 | |
NM_001283047.1 | 5 | 2106 | NP_001269976.1 | |
NM_009594.4 | 4 | 733 | NP_033724.2 | |
AK088252.1 | 5 | 2092 | BAE43394.1 | |
AK090095.1 | 4 | 691 | BAC41088.1 | |
AK166028.1 | 4 | 1951 | ||
AK166210.1 | 5 | 2201 | BAE38629.1 | |
AK219435.1 | 1 | 65 | ||
BC059260.1 | 4 | 760 | AAH59260.1 | |
BC103770.1 | 4 | 1186 | AAI03771.1 | |
CF536749.1 | 2 | 337 | ||
CN663033.1 | 3 | 454 | ||
J02995.1 | 4 | 686 | AAA88241.1 | |
L10656.1 | 2 | 287 |