Assay Details
Target Gene Details
Entrez Gene ID: | 54338 |
Gene Name: | solute carrier family 23 (nucleobase transporters), member 2 |
Gene Aliases: |
AI844736, NBTL1, SVCT2, Slc23a1, YSPL2, YSPL3, mKIAA0238 |
Location: |
Chr.2:132052496-132145108 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc23a2 | NM_018824.2 | 2 | 147 | NP_061294.2 |
AK087175.1 | 2 | 155 | BAC39819.1 | |
AK169991.1 | 2 | 149 | BAE41501.1 | |
AY004874.1 | 2 | 101 | AAG02252.1 |
More Information
Set Membership: |
Intragenic Exonic |