Assay Details
Target Gene Details
Entrez Gene ID: | 15162 |
Gene Name: | hemopoietic cell kinase |
Gene Aliases: |
AI849071, Bmk |
Location: |
Chr.2:153108468-153151441 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hck | NM_001172117.1 | 13 | 1838 | NP_001165588.1 |
NM_010407.4 | 13 | 1838 | NP_034537.2 | |
AI849071.1 | 1 | 270 | ||
AK149736.1 | 13 | 1804 | BAE29054.1 | |
AK150290.1 | 13 | 1812 | BAE29445.1 | |
AK150709.1 | 13 | 1821 | BAE29787.1 | |
AK152506.1 | 13 | 1811 | ||
AK155975.1 | 13 | 1751 | BAE33532.1 | |
AK159234.1 | 13 | 1771 | ||
AK165315.1 | 13 | 1803 | BAE38133.1 | |
BC010478.1 | 13 | 1834 | ||
J03023.1 | 13 | 1727 | AAA37305.1 | |
Y00487.1 | 13 | 1733 | CAA68544.1 |