Assay Details
Target Gene Details
Entrez Gene ID: | 71103 |
Gene Name: | glycosyltransferase 6 domain containing 1 |
Gene Aliases: |
4933411C14Rik, GT6m7 |
Location: |
Chr.2:25771820-25815940 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glt6d1 |
Target Gene Details
Entrez Gene ID: | 620858 |
Gene Name: | predicted gene 13539 |
Gene Aliases: |
OTTMUSG00000012632 |
Location: |
Chr.2:25784617-25787022 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm13539 | NR_045340.1 | 3 | 241 | |
AK076911.1 | 3 | 243 |