Assay Details
Target Gene Details
Entrez Gene ID: | 171469 |
Gene Name: | G protein-coupled receptor 37-like 1 |
Gene Aliases: |
AW047233, CAG-18, D0Kist8 |
Location: |
Chr.1:135160250-135167681 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpr37l1 | NM_134438.3 | 1 | 273 | NP_602320.2 |
AB016602.1 | BAA32062.1 | |||
AK158275.1 | 1 | 275 | ||
BY727284.1 | 1 | 132 |
More Information
Set Membership: |
Intragenic Exonic |