Assay Details
Target Gene Details
Entrez Gene ID: | 103967 |
Gene Name: | dynamin 3 |
Gene Aliases: |
9630020E24Rik, AW061159, B230343F03Rik, C530045C17, mKIAA0820 |
Location: |
Chr.1:161982450-162478323 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnm3 | NM_001038619.1 | 21 | 5121 | NP_001033708.1 |
AK046124.1 | 1 | 657 | ||
AK122379.1 | 12 | 3733 | BAC65661.1 | |
AK147678.1 | 21 | 5065 | BAE28068.1 |
More Information
Set Membership: |
Intragenic Exonic |