Assay Details
Target Gene Details
Entrez Gene ID: | 66052 |
Gene Name: | succinate dehydrogenase complex, subunit C, integral membrane protein |
Gene Aliases: |
0610010E03Rik, AI316496, AU019277 |
Location: |
Chr.1:171129157-171150603 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdhc | NM_025321.3 | 6 | 935 | NP_079597.2 |
AK002459.1 | 6 | 937 | BAB22116.1 | |
AK012818.1 | 6 | 936 | BAB28491.1 | |
AK135503.1 | 6 | 937 | BAE22557.1 | |
AV356234.1 | ||||
BC005779.1 | 6 | 922 | AAH05779.1 | |
BC083091.1 | 6 | 928 | AAH83091.1 | |
BY711932.1 |
More Information
Set Membership: |
Intragenic Exonic |