Assay Details
Target Gene Details
Entrez Gene ID: | 66208 |
Gene Name: | neuron derived neurotrophic factor |
Gene Aliases: |
1110060M21Rik, SCIRP10, Spuf |
Location: |
Chr.1:191306797-191318118 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nenf | NM_025424.2 | 4 | 559 | NP_079700.1 |
AB126218.1 | BAD72062.1 | |||
AK002413.1 | 4 | 560 | BAB22081.1 | |
AK004329.1 | 4 | 548 | BAB23264.1 | |
AK009304.1 | 4 | 550 | BAB26205.1 | |
AK020403.1 | 4 | 541 | BAB32092.1 | |
AK155319.1 | 4 | 558 | BAE33187.1 | |
BC048464.1 | 4 | 547 | AAH48464.1 |
More Information
Set Membership: |
Intragenic Exonic |