Assay Details
Target Gene Details
Entrez Gene ID: | 269152 |
Gene Name: | kinesin family member 26B |
Gene Aliases: |
4832420M10, BC056349, D230039L06Rik |
Location: |
Chr.1:178525741-178939200 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kif26b | NM_001161665.1 | 2 | 833 | NP_001155137.1 |
AB355846.1 | 2 | 833 | ||
AK052046.1 | 2 | 834 | BAC34839.1 | |
BC120791.1 | 2 | 496 | ||
BC137689.1 | 2 | 496 |
More Information
Set Membership: |
Intragenic Exonic |