Assay Details
Target Gene Details
Entrez Gene ID: | 16151 |
Gene Name: | inhibitor of kappaB kinase gamma |
Gene Aliases: |
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO |
Location: |
Chr.X:74393291-74456792 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ikbkg | NM_001136067.2 | 9 | 1222 | NP_001129539.1 |
NM_001161421.1 | 9 | 1219 | NP_001154893.1 | |
NM_001161422.1 | 8 | 1040 | NP_001154894.1 | |
NM_001161423.1 | 8 | 1092 | NP_001154895.1 | |
NM_001161424.1 | 8 | 1089 | NP_001154896.1 | |
NM_010547.2 | 8 | 1118 | NP_034677.2 | |
NM_178590.4 | 8 | 1115 | NP_848705.1 | |
AF069542.1 | 7 | 977 | AAC40153.1 | |
AF513109.1 | 9 | 1133 | AAP47160.1 | |
AK037020.1 | 8 | 1094 | BAC29672.1 | |
AK154095.1 | 8 | 1104 | BAE32372.1 | |
AY112937.1 | 9 | 1222 | AAM54725.1 | |
BC021431.1 | 9 | 1209 | AAH21431.1 |