Assay Details
Target Gene Details
Entrez Gene ID: | 245533 |
Gene Name: | acyl-CoA wax alcohol acyltransferase 1 |
Gene Aliases: |
Dgat2l3 |
Location: |
Chr.X:100572247-100578206 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Awat1 | NM_001081136.1 | NP_001074605.1 | ||
AK137763.1 | ||||
BC147680.1 | ||||
BC147683.1 | ||||
BC147829.1 | ||||
BC147832.1 |
More Information
Set Membership: |
Intragenic Non-exonic |