Assay Details
Target Gene Details
Entrez Gene ID: | 68117 |
Gene Name: | apolipoprotein O-like |
Gene Aliases: |
6720473G16Rik, 9430083G14Rik, E130106L15Rik |
Location: |
Chr.X:112311408-112372755 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apool | NM_026565.2 | NP_080841.1 | ||
AK012398.1 | ||||
AK012804.1 | BAB28482.1 | |||
AK020156.1 | ||||
AK028953.1 | ||||
AK079141.1 | ||||
AK086522.1 | ||||
AK088869.1 | ||||
BC024334.1 | AAH24334.1 |