Assay Details
Target Gene Details
Entrez Gene ID: | 17995 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 1 |
Gene Aliases: |
CI-51kD |
Location: |
Chr.19:4007499-4012755 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufv1 | NM_133666.3 | 4 | 595 | NP_598427.1 |
AK005021.1 | 4 | 545 | ||
AK075692.1 | 4 | 540 | BAC35893.1 | |
AK151455.1 | 4 | 506 | ||
AK187072.1 | ||||
AK189391.1 | ||||
AK206448.1 | ||||
AK211390.1 | ||||
AK218269.1 | ||||
BC014818.1 | 4 | 565 | AAH14818.1 | |
BC041682.1 | 4 | 498 | AAH41682.1 |
More Information
Set Membership: |
Intragenic Exonic |