Assay Details
Target Gene Details
Entrez Gene ID: | 20510 |
Gene Name: | solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1 |
Gene Aliases: |
D130048G10Rik, EAAC1, EAAC2, EAAT3, MEAAC1 |
Location: |
Chr.19:28835166-28913960 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc1a1 | NM_009199.2 | 1 | 43 | NP_033225.1 |
AK018188.1 | 1 | 76 | BAB31114.1 | |
AK045632.1 | 1 | 75 | ||
AK051428.1 | 1 | 75 | ||
AK083097.1 | 1 | 75 | ||
BC064797.1 | 1 | 65 | AAH64797.1 | |
BC065099.1 | 1 | 67 | AAH65099.1 | |
D43797.1 | 1 | 49 | BAA07855.1 | |
U73521.1 | 1 | 43 | AAB80694.1 |
More Information
Set Membership: |
Intragenic Exonic |