Assay Details
Target Gene Details
Entrez Gene ID: | 11632 |
Gene Name: | aryl-hydrocarbon receptor-interacting protein |
Gene Aliases: |
AA408703, AW476050, Ara9, D19Bwg1412e, Fkbp16, Xap2 |
Location: |
Chr.19:4113756-4127236 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aip |
Target Gene Details
Entrez Gene ID: | 66990 |
Gene Name: | transmembrane protein 134 |
Gene Aliases: |
2410001H17Rik, AI463452 |
Location: |
Chr.19:4125960-4132307 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem134 | NM_001078649.1 | 1 | 165 | NP_001072117.1 |
NM_025889.2 | 1 | 165 | NP_080165.1 | |
AK002220.1 | 1 | 159 | BAB21944.1 | |
AK008515.1 | 1 | 139 | BAB25713.1 | |
AK009263.1 | 1 | 167 | ||
AK009745.1 | 1 | 141 | BAB26475.1 | |
AK010323.1 | 1 | 167 | BAC25290.1 | |
AK129014.1 | 1 | 138 | BAC87670.1 | |
AK166519.1 | 1 | 168 | BAE38823.1 | |
BC026765.1 | 1 | 147 | AAH26765.1 | |
CF983315.1 | 1 | 191 |