Assay Details
Target Gene Details
Entrez Gene ID: | 14682 |
Gene Name: | guanine nucleotide binding protein, alpha q polypeptide |
Gene Aliases: |
1110005L02Rik, 6230401I02Rik, AA408290, AW060788, Dsk1, Dsk10, Galphaq, Gq, GqI |
Location: |
Chr.19:16132831-16387453 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnaq | NM_008139.5 | 3 | 851 | NP_032165.3 |
AK085933.1 | ||||
AK147393.1 | 3 | 852 | BAE27882.1 | |
BC057583.1 | 3 | 591 | AAH57583.1 | |
M55412.1 | 3 | 392 | AAA63306.1 |
More Information
Set Membership: |
Intragenic Exonic |