Assay Details
Target Gene Details
Entrez Gene ID: | 12460 |
Gene Name: | copper chaperone for superoxide dismutase |
Gene Aliases: |
Ccsd |
Location: |
Chr.19:4825366-4839327 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccs | NM_016892.3 | 3 | 275 | NP_058588.1 |
AF121906.1 | 3 | 188 | AAD23832.1 | |
AF173379.1 | 3 | 248 | AAF70242.1 | |
AK050072.1 | 3 | 224 | BAC34057.1 | |
BC026938.1 | 3 | 242 | AAH26938.1 | |
BY073478.1 | 3 | 275 |
More Information
Set Membership: |
Intragenic Exonic |