Assay Details
Target Gene Details
Entrez Gene ID: | 66388 |
Gene Name: | cutC copper transporter homolog (E.coli) |
Gene Aliases: |
2310039I18Rik, AI326282, CGI-32 |
Location: |
Chr.19:43753023-43768638 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cutc | NM_001113562.1 | 6 | 662 | NP_001107034.1 |
NM_025530.3 | 6 | 632 | NP_079806.2 | |
AK007591.1 | 6 | 587 | BAB25124.1 | |
AK009702.1 | 6 | 657 | BAB26449.1 | |
BC026775.1 | 6 | 572 | AAH26775.1 |
More Information
Set Membership: |
Intragenic Exonic |