Assay Details
Target Gene Details
Entrez Gene ID: | 18145 |
Gene Name: | Niemann-Pick type C1 |
Gene Aliases: |
A430089E03Rik, C85354, D18Ertd139e, D18Ertd723e, lcsd, nmf164, spm |
Location: |
Chr.18:12189694-12236386 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npc1 | NM_008720.2 | 4 | 518 | NP_032746.2 |
AF003348.1 | 4 | 495 | AAB63372.1 | |
AK044878.1 | 3 | 2911 | BAC32127.1 | |
AK087174.1 | 3 | 2925 | ||
AK154282.1 | 4 | 438 | BAE32486.1 | |
AK155379.1 | 4 | 438 | BAE33230.1 | |
AK170880.1 | 3 | 2911 | ||
AK171371.1 | 2 | 4031 | ||
BC052437.1 | 4 | 518 | AAH52437.1 | |
BC054539.1 | 4 | 518 | AAH54539.1 |