Assay Details
Target Gene Details
Entrez Gene ID: | 72432 |
Gene Name: | serine peptidase inhibitor, Kazal type 5 |
Gene Aliases: |
2310065D10Rik, AV238912, LEKT1, LETK1, VAKT1 |
Location: |
Chr.18:43959545-44022487 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spink5 | NM_001081180.1 | 3 | 208 | NP_001074649.1 |
AJ426449.2 | 3 | 206 | CAD20045.2 | |
AK035690.1 | 3 | 209 | BAC29153.1 | |
AK036804.1 | 3 | 209 | BAC29581.1 | |
AK162690.1 | 3 | 209 | BAE37024.1 | |
BC118011.1 | 3 | 180 |
More Information
Set Membership: |
Intragenic Exonic |