Assay Details
Target Gene Details
Entrez Gene ID: | 107035 |
Gene Name: | F-box protein 38 |
Gene Aliases: |
6030410I24Rik, AU044865, AW214031, Moka, SP329 |
Location: |
Chr.18:62504059-62548858 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbxo38 | NM_134136.3 | 14 | 2041 | NP_598897.2 |
AK031096.1 | 14 | 1991 | BAC27250.1 | |
AK031347.1 | 14 | 1972 | BAC27358.1 | |
AK034761.1 | 9 | 1225 | BAC28826.1 | |
AK089046.1 | 14 | 2013 | BAC40718.1 | |
AY267463.1 | 14 | 1974 | AAP31028.1 | |
BC056348.1 | 14 | 1993 | AAH56348.1 |