Assay Details
Target Gene Details
Entrez Gene ID: | 108077 |
Gene Name: | superkiller viralicidic activity 2-like (S. cerevisiae) |
Gene Aliases: |
4930534J06Rik, AW214248, Ddx13, SKI, Ski2w |
Location: |
Chr.17:34839226-34850204 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Skiv2l | NM_021337.2 | 19 | 2318 | NP_067312.2 |
AK029763.1 | 19 | 2307 | BAC26604.1 | |
AK087810.1 | 15 | 2951 | BAC40010.1 | |
AK139051.1 | 18 | 2392 | ||
AK159854.1 | 19 | 2319 | BAE35430.1 | |
AK169864.1 | 19 | 2289 | BAE41420.1 | |
AK180219.1 | ||||
AK185511.1 | ||||
AK214632.1 | ||||
BC023478.1 | 7 | 922 | AAH23478.1 | |
BC065999.1 | 19 | 2290 | AAH65999.1 |
More Information
Set Membership: |
Intragenic Exonic |