Assay Details
Target Gene Details
Entrez Gene ID: | 76482 |
Gene Name: | RIKEN cDNA 3110002H16 gene |
Gene Aliases: |
2400010D15Rik, Mic-1, Mic1 |
Location: |
Chr.18:12168691-12198497 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
3110002H16Rik | NM_029623.2 | NP_083899.1 | ||
AK010309.1 | ||||
AK144623.1 | BAE25975.1 | |||
AK154240.1 | BAE32456.1 | |||
AK159691.1 | BAE35292.1 | |||
BC021846.1 | AAH21846.1 |