Assay Details
Target Gene Details
Entrez Gene ID: | 76482 |
Gene Name: | RIKEN cDNA 3110002H16 gene |
Gene Aliases: |
2400010D15Rik, Mic-1, Mic1 |
Location: |
Chr.18:12168691-12198497 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
3110002H16Rik | NM_029623.2 | 15 | 1423 | NP_083899.1 |
AF003352.1 | 3 | 309 | AAB63375.1 | |
AK013979.1 | 9 | 819 | ||
AK144623.1 | 15 | 1424 | BAE25975.1 | |
AK154240.1 | 15 | 1418 | BAE32456.1 | |
AK159691.1 | 15 | 1418 | BAE35292.1 | |
BC021846.1 | 15 | 1406 | AAH21846.1 |