Assay Details
Target Gene Details
Entrez Gene ID: | 20532 |
Gene Name: | solute carrier family 3, member 1 |
Gene Aliases: |
D2H, NBAT, NTAA, RBAT |
Location: |
Chr.17:85028347-85064241 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc3a1 | NM_009205.2 | NP_033231.2 | ||
AK052623.1 | BAC35066.1 | |||
AK132275.1 | BAE21075.1 | |||
BC013441.1 | AAH13441.1 | |||
D88533.1 | BAA24427.1 |
More Information
Set Membership: |
Intragenic Non-exonic |