Assay Details
Target Gene Details
Entrez Gene ID: | 116871 |
Gene Name: | metastasis associated 3 |
Gene Aliases: |
1110002J22Rik, mKIAA1266 |
Location: |
Chr.17:83706163-83823715 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mta3 | NM_001171052.1 | NP_001164523.1 | ||
NM_001171053.1 | NP_001164524.1 | |||
NM_001171054.1 | NP_001164525.1 | |||
NM_054082.2 | NP_473423.1 | |||
AF288138.1 | AAK83045.1 | |||
AK129318.1 | BAC98128.1 | |||
AK145942.1 | BAE26772.1 | |||
AK146721.1 | BAE27384.1 | |||
AK146905.1 | BAE27518.1 | |||
AK147197.1 | BAE27755.1 | |||
AK154856.1 | BAE32881.1 | |||
AK158866.1 | BAE34701.1 | |||
BC022124.1 | AAH22124.1 | |||
BC099960.1 | ||||
BU756693.1 | ||||
CA751287.1 |
More Information
Set Membership: |
Intragenic Non-exonic |