Assay Details
Target Gene Details
Entrez Gene ID: | 69875 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11 |
Gene Aliases: |
2010012C24Rik, AV006275, B14.7 |
Location: |
Chr.17:56717762-56724248 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufa11 | NM_027244.1 | NP_081520.1 | ||
AK008201.1 | BAB25529.1 | |||
BC059729.1 | AAH59729.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |