Assay Details
Target Gene Details
Entrez Gene ID: | 109801 |
Gene Name: | glyoxalase 1 |
Gene Aliases: |
0610009E22Rik, 1110008E19Rik, 2510049H23Rik, AW550643, GLY1, Glo-1, Glo-1r, Glo-1s, Glo1-r, Glo1-s, Qglo |
Location: |
Chr.17:30592861-30612659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glo1 | NM_001113560.1 | NP_001107032.1 | ||
NM_025374.3 | NP_079650.3 | |||
AK002386.1 | BAB22060.1 | |||
AK003567.1 | BAB22863.1 | |||
AK005055.1 | BAB23781.1 | |||
AK005215.1 | ||||
AK031832.1 | BAC27570.1 | |||
AK049703.1 | BAC33882.1 | |||
BC024663.1 | AAH24663.1 | |||
BC081432.1 | AAH81432.1 | |||
BG077218.2 | ||||
BY312309.1 | ||||
DQ487161.1 | ||||
DQ487162.1 |
More Information
Set Membership: |
Intragenic Non-exonic |