Assay Details
Target Gene Details
Entrez Gene ID: | 107766 |
Gene Name: | 3-hydroxyanthranilate 3,4-dioxygenase |
Gene Aliases: |
0610007K21Rik, 0610012J07Rik, 3-HAO, 3-HAOxase, 3HAO |
Location: |
Chr.17:83831328-83847950 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Haao | NM_025325.2 | NP_079601.1 | ||
AK002295.1 | ||||
AK002608.1 | ||||
AK003233.1 | ||||
BC012872.1 | AAH12872.1 | |||
BC094021.1 | AAH94021.1 |
More Information
Set Membership: |
Intragenic Non-exonic |