Assay Details
Target Gene Details
Entrez Gene ID: | 64337 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 13 |
Gene Aliases: |
1500031D04Rik, AB030194, Ggamma13 |
Location: |
Chr.17:25717172-25719102 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gng13 | NM_022422.5 | NP_071867.1 | ||
AB030194.1 | BAA92757.1 | |||
AK018778.1 | BAB31403.1 | |||
AY029485.1 | AAK40268.1 | |||
BC048431.1 | AAH48431.1 | |||
BC059712.1 | AAH59712.1 | |||
BU554843.1 |
More Information
Set Membership: |
Intragenic Non-exonic |